missing translation for 'onlineSavingsMsg'
Få mere at vide

2-(4-(Methylsulfonamido)phenyl)acetic Acid, TRC

Artikelnummer. 30466884
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30466884 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30466884 Leverandør Toronto Research Chemicals Leverandørnr. TRCM3322201G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale 2-(4-(Methylsulfonamido)phenyl)acetic Acid
CAS 56205-88-0
Molekylær formel C9H11NO4S
Mængde 1 g
Synonym 4-(Methylsulfonylamino)phenylacetic Acid, 4-Methanesulfonamidobenzeneacetic Acid, UK 66544, 4-[(Methylsulfonyl)amino]benzeneacetic Acid
InChI formel InChI=1S/C9H11NO4S/c1-15(13,14)10-8-4-2-7(3-5-8)6-9(11)12/h2-5,10H,6H2,1H3,(H,11,12)
SMIL CS(=O)(=O)NC1=CC=C(C=C1)CC(=O)O
IUPAC navn 2-[4-(methanesulfonamido)phenyl]acetic acid
Molekylvægt (g/mol) 229.25
Formel vægt 229.0409
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.