Få mere at vide
2-(4-methylphenyl)oxirane, TRC
Artikelnummer. 30473109
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30473109 100 mg
30464962 10 mg
30471232 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30473109 Leverandør Toronto Research Chemicals Leverandørnr. TRCM355488100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 13107-39-6
Molekylær formel C9H10O
Molekylvægt (g/mol) 134.17
IUPAC navn 2-(4-methylphenyl)oxirane
SMIL Cc1ccc(cc1)C2CO2
Tekniske data
Kemisk navn eller materiale 2-(4-methylphenyl)oxirane
Mængde 100 mg
InChI formel InChI=1S/C9H10O/c1-7-2-4-8(5-3-7)9-6-10-9/h2-5,9H,6H2,1H3
Formel vægt 134.0732