missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(4-Methyl-6,7-dihydro-4H-thieno[3,2-c]pyran-4-yl)ethanol, 95%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 94.5 |
CAS Max % | 100.0 |
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
IUPAC Name | 2-{4-methyl-4H,6H,7H-thieno[3,2-c]pyran-4-yl}ethan-1-ol |
Formula Weight | 198.28 |
Percent Purity | 95% |
Chemical Name or Material | 2-(4-Methyl-6, 7-dihydro-4H-thieno[3, 2-c]pyran-4-yl)ethanol |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'