Få mere at vide
2-(4-Methyl-2-phenyl-1-piperazinyl)-3-pyridinemethanol, TRC
Artikelnummer. 30138644
Skift visning
Mængde:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30138644 250 mg
17902925 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30138644 Leverandør Toronto Research Chemicals Leverandørnr. TRCM320640250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 61337-89-1
Molekylær formel C17 H21 N3 O
Molekylvægt (g/mol) 283.37
Synonym [2-[(2RS)-4-Methyl-2-phenylpiperazin-1-yl]pyridin-3-yl]methanol, 3-Pyridinemethanol, 2-(4-methyl-2-phenyl-1-piperazinyl)-, 2-(4-Methyl-2-phenyl-1-piperazinyl)-3-pyridinemethanol, 1-(3-Hydroxymethyl-2-pyridyl)-4-methyl-2-phenylpiperazine, 1-[3-(Hydroxymethyl)pyridin-2-yl)-4-methyl-2-phenylpiperazine, 2-(4-Methyl-2-phenylpiperazin-1-yl)pyridine-3-methanol
IUPAC navn [2-(4-methyl-2-phenylpiperazin-1-yl)pyridin-3-yl]methanol
SMIL CN1CCN(C(C1)c2ccccc2)c3ncccc3CO
Tekniske data
Kemisk navn eller materiale (2-[(2RS)-4-Methyl-2-phenyl-piperazin-1-yl]pyridin-3-yl)methanol
Mængde 250 mg
InChI formel InChI=1S/C17H21N3O/c1-19-10-11-20(17-15(13-21)8-5-9-18-17)16(12-19)14-6-3-2-4-7-14/h2-9,16,21H,10-13H2,1H3
Formel vægt 283.1685