missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Methyl-1-piperazinyl)thiazole-5-carboxaldehyde, 99%, Thermo Scientific™

Catalog Number p-7033364
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
320423-50-5
C9H13N3OS
211.283
MFCD01568852
TUJAFVJUJXMFEG-UHFFFAOYSA-N
2-4-methylpiperazin-1-yl-1,3-thiazole-5-carbaldehyde, 2-4-methylpiperazino-1,3-thiazole-5-carbaldehyde, 2-4-methylpiperazin-1-yl thiazole-5-carbaldehyde, 2-4-methyl-1-piperazinyl thiazole-5-carboxaldehyde, 2-4-methylpiperazin-1-yl thiazole-5-carboxaldehyde, 2-4-methylpiperazinyl-1,3-thiazole-5-carbaldehyde, acmc-20aoov, methylpiperazinothiazolecarbaldehyde
1490234
2-(4-methylpiperazin-1-yl)-1,3-thiazole-5-carbaldehyde
CN1CCN(CC1)C2=NC=C(S2)C=O
This item is not returnable. View return policy
320423-50-5
C9H13N3OS
211.283
MFCD01568852
TUJAFVJUJXMFEG-UHFFFAOYSA-N
2-4-methylpiperazin-1-yl-1,3-thiazole-5-carbaldehyde, 2-4-methylpiperazino-1,3-thiazole-5-carbaldehyde, 2-4-methylpiperazin-1-yl thiazole-5-carbaldehyde, 2-4-methyl-1-piperazinyl thiazole-5-carboxaldehyde, 2-4-methylpiperazin-1-yl thiazole-5-carboxaldehyde, 2-4-methylpiperazinyl-1,3-thiazole-5-carbaldehyde, acmc-20aoov, methylpiperazinothiazolecarbaldehyde
1490234
2-(4-methylpiperazin-1-yl)-1,3-thiazole-5-carbaldehyde
CN1CCN(CC1)C2=NC=C(S2)C=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 211.29
Percent Purity 99%
Chemical Name or Material 2-(4-Methyl-1-piperazinyl)thiazole-5-carboxaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'