Få mere at vide
2-(4-Methoxyphenyl)-N,N-dimethylethanamine Hydrochloride, Mikromol™
Beskrivelse
2-(4-Methoxyphenyl)-N,N-dimethylethanamine Hydrochloride
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 2-(4-Methoxyphenyl)-N,N-dimethylethanamine Hydrochloride |
| CAS | 50822-98-5 |
| Analyt- eller komponentnavne | 2-(4-Methoxyphenyl)-N,N-dimethylethanamine Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C11 H17 N O . Cl H |
| Certificeringer/compliance | ISO 17025 |
| Synonym | Venlafaxine Hydrochloride Imp. A (EP), Venlafaxine Imp. A (EP), 2-(4-Methoxyphenyl)-N,N-dimethylethanamine Hydrochloride, Venlafaxine Hydrochloride Imp. A (EP) as Hydrochloride, Venlafaxine Hydrochloride Impurity A as Hydrochloride, Venlafaxine Impurity A as Hydrochloride |
| InChI formel | InChI=1S/C11H17NO.ClH/c1-12(2)9-8-10-4-6-11(13-3)7-5-10;/h4-7H,8-9H2,1-3H3;1H |
| SMIL | Cl.COc1ccc(CCN(C)C)cc1 |
| IUPAC navn | 2-(4-methoxyphenyl)-N,N-dimethylethanamine;hydrochloride |
| Vis mere |