Få mere at vide
2-[[(4-Methoxy-3,5-dimethylpyridin-2-yl)-methyl]sulphinyl]-1H-benzimidazole, Mikromol™
Artikelnummer. 30051230
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30051230 100 mg
30052147 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30051230 Leverandør Mikromol Leverandørnr. MM0095.23
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2-[[(4-Methoxy-3,5-dimethylpyridin-2-yl)-methyl]sulphinyl]-1H-benzimidazole
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-[((4-Methoxy-3,5-dimethylpyridin-2-yl)-methyl)sulphinyl]-1H-benzimidazole
CAS 73590-60-0
Analyt- eller komponentnavne 2-[((4-Methoxy-3,5-dimethylpyridin-2-yl)-methyl)sulphinyl]-1H-benzimidazole
Forsendelsestilstand Ice pack (-20°C)
Molekylær formel C16 H17 N3 O2 S
Certificeringer/compliance ISO 17025
Synonym 2-[[(4-Methoxy-3,5-dimethylpyridin-2-yl)-methyl]sulphinyl]-1H-benzimidazole
InChI formel InChI=1S/C16H17N3O2S/c1-10-8-17-14(11(2)15(10)21-3)9-22(20)16-18-12-6-4-5-7-13(12)19-16/h4-8H,9H2,1-3H3,(H,18,19)
SMIL COc1c(C)cnc(CS(=O)c2nc3ccccc3[nH]2)c1C
IUPAC navn 2-[(4-methoxy-3,5-dimethylpyridin-2-yl)methylsulfinyl]-1H-benzimidazole
Molekylvægt (g/mol) 315.39
Mængde 100 mg
Formel vægt 315.1041 g/mol