Få mere at vide
2-(4-Isobutyrylphenyl)propanamide, Mikromol™
Artikelnummer. 30460615
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30460615 4 x 25 mg
17900506 100 mg
17990936 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30460615 Leverandør Mikromol Leverandørnr. MM0002.0700254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2-(4-Isobutyrylphenyl)propanamide, Mikromol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-(4-Isobutyrylphenyl)propanamide
CAS 1823308-53-7
Analyt- eller komponentnavne 2-(4-Isobutyrylphenyl)propanamide
Forsendelsestilstand Room Temperature
Molekylær formel C13 H17 N O2
Certificeringer/compliance ISO 17025
Synonym 2-(4-Isobutyrylphenyl)propanamide
InChI formel InChI=1S/C13H17NO2/c1-8(2)12(15)11-6-4-10(5-7-11)9(3)13(14)16/h4-9H,1-3H3,(H2,14,16)
Løsningstype Neat
SMIL CC(C)C(=O)c1ccc(cc1)C(C)C(=O)N
IUPAC navn 2-[4-(2-methylpropanoyl)phenyl]propanamide
Molekylvægt (g/mol) 219.28
Mængde 4 x 25 mg
Formel vægt 219.1259 g/mol