Få mere at vide
2-(4-Hydroxyphenoxy-2,3,5,6-d4)-propanoic Acid, TRC
Artikelnummer. 30408757
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30408757 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30408757 Leverandør Toronto Research Chemicals Leverandørnr. TRCH97292710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-(4-Hydroxyphenoxy)propanoic Acid D4 (phenoxy-2,3,5,6-D4)
CAS 127893-32-7
Molekylær formel C9 2H4 H6 O4
Mængde 10 mg
Synonym Propanoic acid, 2-(4-hydroxyphenoxy-2,3,5,6-d4)- (9CI), 2-(4-Hydroxyphenoxy)propanoic acid D4 (phenoxy-2,3,5,6-D4), 2-(4-Hydroxyphenoxy-2,3,5,6-d4)propanoic acid, (±)-2-(4-Hydroxyphenoxy-2,3,5,6-d4)propionic acid, O-(4-Hydroxyphenyl-d4)lactic acid, α-(p-Hydroxyphenoxy-d4)propionic acid
InChI formel InChI=1S/C9H10O4/c1-6(9(11)12)13-8-4-2-7(10)3-5-8/h2-6,10H,1H3,(H,11,12)/i2D,3D,4D,5D
SMIL [2H]c1c([2H])c(OC(C)C(=O)O)c([2H])c([2H])c1O
IUPAC navn 2-(2,3,5,6-tetradeuterio-4-hydroxyphenoxy)propanoic acid
Molekylvægt (g/mol) 186.198
Formel vægt 186.083