Få mere at vide
2-(4-Hydroxy-3-methoxyphenyl)ethyl-1,1,2,2-d4-amine HCl, CDN
Artikelnummer. 30318134
Skift visning
Mængde:
0.01 g
0.05 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30318134 0.01 g
30316141 0.05 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30318134 Leverandør CDN Leverandørnr. CDND22100.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3-Methoxy Dopamine-d4 Hydrochloride
CAS 1216788-76-9
Molekylær formel C9 2H4 H9 N O2 . Cl H
Mængde 0.01 g
InChI formel InChI=1 S/C9H13NO2.ClH/c1-12-9-6-7(4-5-10)2-3-8(9)11;/h2-3,6,11 H,4-5,10H2,1H3;1 H/i4D2,5D2;
SMIL Cl.[2 H]C([2 H])(N)C([2 H])([2 H])c1ccc(O)c(OC)c1
IUPAC navn 4-(2-amino-1,1,2,2-tetradeuterioethyl)-2-methoxyphenol;hydrochloride
Molekylvægt (g/mol) 207.69
Formel vægt 207.0964 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity