Få mere at vide
2-(4-Fluorophenoxy-d4)-N-methoxy-N-methyl-1-butene, TRC
Artikelnummer. 30408591
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30408591 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30408591 Leverandør Toronto Research Chemicals Leverandørnr. TRCF595552100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-(4-Fluorophenoxy-d4)-N-methoxy-N-methyl-1-butene
CAS 1346606-16-3
Molekylær formel C12H10D4FNO3
Mængde 100 mg
InChI formel InChI=1S/C12H14FNO3/c1-14(16-2)8-7-11(15)9-17-12-5-3-10(13)4-6-12/h3-8H,9H2,1-2H3/b8-7+/i3D,4D,5D,6D
SMIL [2H]c1c([2H])c(OCC(=O)\C=C\N(C)OC)c([2H])c([2H])c1F
IUPAC navn (E)-4-[methoxy(methyl)amino]-1-(2,3,5,6-tetradeuterio-4-fluorophenoxy)but-3-en-2-one
Molekylvægt (g/mol) 243.27
Formel vægt 243.1209