Learn More
2'-(4-Ethoxyphenyl)-5-(4-methylpiperazin-1-yl)-1H,1'H-2,5'-bibenzo[d]imidazole, 98%, BLD Pharm™
Product Code. 30947207
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30947207 250 mg 250mg
30943494 100 mg 100mg
30943319 1 g 1g
30939340 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 30947207 Supplier BLD Pharm Supplier No. BD133718250mg
 Restricted Item
Forventet forsendelse: 27-11-2026
Specifications
CAS 23491-52-3
Molecular Formula C27H28N6O
MDL Number MFCD00166996
Quantity 250 mg
SMILES CN1CCN(C2=CC=C3NC(C4=CC=C5NC(C6=CC=C(OCC)C=C6)=NC5=C4)=NC3=C2)CC1
Molecular Weight (g/mol) 452.55
Percent Purity 98%
Chemical Name or Material 2'-(4-Ethoxyphenyl)-5-(4-methylpiperazin-1-yl)-1H,1'H-2,5'-bibenzo[d]imidazole