Få mere at vide
2,4-Dithiouridine, TRC
Artikelnummer. 30378392
Skift visning
Mængde:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30378392 250 mg
30375105 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30378392 Leverandør Toronto Research Chemicals Leverandørnr. TRCD494370250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 13239-96-8
Molekylær formel C9 H12 N2 O4 S2
Molekylvægt (g/mol) 276.33
IUPAC navn 1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dithione
SMIL OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N2C=CC(=S)NC2=S
Tekniske data
Kemisk navn eller materiale 2,4-Dithiouridine
Mængde 250 mg
InChI formel InChI=1S/C9H12N2O4S2/c12-3-4-6(13)7(14)8(15-4)11-2-1-5(16)10-9(11)17/h1-2,4,6-8,12-14H,3H2,(H,10,16,17)/t4-,6-,7-,8-/m1/s1
Formel vægt 276.0238