Få mere at vide
(2,4-Dioxo-thiazolidin-3-yl)acetic Acid, TRC
Artikelnummer. 30180177
Skift visning
Mængde:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30180177 500 mg
30179966 100 mg
30179919 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30180177 Leverandør Toronto Research Chemicals Leverandørnr. TRCD487080500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 31061-24-2
Molekylær formel C5H5NO4S
Molekylvægt (g/mol) 175.16
Synonym (2,4-Dioxo-thiazolidin-3-yl)-acetic acid
SMIL OC(=O)CN1C(=O)CSC1=O
Tekniske data
Kemisk navn eller materiale (2,4-Dioxo-thiazolidin-3-yl)acetic Acid
Mængde 500 mg
InChI formel InChI=1S/C5H5NO4S/c7-3-2-11-5(10)6(3)1-4(8)9/h1-2H2,(H,8,9)
Formel vægt 174.9939