Få mere at vide
2,4-Dinitrophenol-d3 (wetted with >15% H2O), TRC
Artikelnummer. 30372624
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30372624 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30372624 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4799475MG
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2,4-Dinitrophenol D3
CAS 93951-77-0
Renhedsgrad noter HPLC
Molekylær formel C6 D3 H N2 O5
Mængde 5 mg
Synonym Phen-2,3,5-d3-ol, 4,6-dinitro- (9CI), 2,4-Dinitrophenol D3, 2,4-Dinitrophenol (ring-D3), 2,4-Dinitrophenol-3,5,6-D3, 2,4-Dinitro-3,5,6-trideuteriophenol
InChI formel InChI=1S/C6H4N2O5/c9-6-2-1-4(7(10)11)3-5(6)8(12)13/h1-3,9H/i1D,2D,3D
SMIL [2H]c1c([2H])c(c([2H])c(c1O)[N+](=O)[O-])[N+](=O)[O-]
IUPAC navn 2,3,5-trideuterio-4,6-dinitro-phenol
Molekylvægt (g/mol) 187.125
Formel vægt 187.0309
Procent renhed >95