missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethylimidazole, 90%, Tech., Thermo Scientific Chemicals

Catalog Number p-118046
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
930-62-1
C5H8N2
96.13
MFCD00022365
LLPKQRMDOFYSGZ-UHFFFAOYSA-N
2,4-dimethylimidazole, 2,4-dimethyl-1h-imidazole, 1h-imidazole, 2,4-dimethyl, 2,4-dimethyl-3h-imidazole, 2,4-dimethyl-1h-imidazol, imidazole, 2,4-dimethyl, 2,4-dimethyl, 2,5-dimethylimidazole, imidazole,4-dimethyl, pubchem18587
70259
2,5-dimethyl-1H-imidazole
CC1=CN=C(C)N1
This item is not returnable. View return policy
930-62-1
C5H8N2
96.13
MFCD00022365
LLPKQRMDOFYSGZ-UHFFFAOYSA-N
2,4-dimethylimidazole, 2,4-dimethyl-1h-imidazole, 1h-imidazole, 2,4-dimethyl, 2,4-dimethyl-3h-imidazole, 2,4-dimethyl-1h-imidazol, imidazole, 2,4-dimethyl, 2,4-dimethyl, 2,5-dimethylimidazole, imidazole,4-dimethyl, pubchem18587
70259
2,5-dimethyl-1H-imidazole
CC1=CN=C(C)N1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 88.0
CAS Max % 100.0
Melting Point 85.0°C to 87.0°C
Color Yellow to Orange
Boiling Point 266.0°C (733.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Quantity 5 g
Beilstein 23, V,5, 172
Formula Weight 96.13
Percent Purity 90%
Grade Technical
Physical Form Viscous Liquid or Weak Mass
Chemical Name or Material 2, 4-Dimethylimidazole, tech., 90%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,4-Dimethylimidazole
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'