Få mere at vide
2,4-Dimethyl-d6-aniline HCl, CDN
Artikelnummer. 30314596
Skift visning
Mængde:
0.1 g
0.25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30314596 0.1 g
30314574 0.25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30314596 Leverandør CDN Leverandørnr. CDND56840.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2,4-Dimethylaniline-D6 (dimethyl-D6) Hydrochloride
CAS 1219805-38-5
Molekylær formel C8 D6 H5 N . Cl H
Mængde 0.1 g
Synonym 2,4-Dimethylaniline-D6 (dimethyl-D6) hydrochloride, 2,4-Di(methyl-d3)benzenamine hydrochloride, 2,4-Xylidine-d6 hydrochloride, 2,4-Di(methyl-d3)aniline hydrochloride
InChI formel InChI=1 S/C8H11N.ClH/c1-6-3-4-8(9)7(2)5-6;/h3-5 H,9H2,1-2H3;1 H/i1D3,2D3;
SMIL Cl.[2 H]C([2 H])([2 H])c1ccc(N)c(c1)C([2 H])([2 H])[2 H]
IUPAC navn 2,4-bis(trideuteriomethyl)aniline;hydrochloride
Molekylvægt (g/mol) 163.6776
Formel vægt 163.1035 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity