missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Dimethoxybenzaldehyde, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
2,4-Dimethoxybenzaldehyde is used as a reagent to specifically quantify phlorotannins. It is also used as a component of hair dyes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,4-Dimethoxybenzaldehyde is used as a reagent to specifically quantify phlorotannins. It is also used as a component of hair dyes.
Solubility
Insoluble in water. Soluble in chlorofrom.
Notes
Air sensitive. Incompatible with oxidizing agents.
2,4-Dimethoxybenzaldehyde is used as a reagent to specifically quantify phlorotannins. It is also used as a component of hair dyes.
Solubility
Insoluble in water. Soluble in chlorofrom.
Notes
Air sensitive. Incompatible with oxidizing agents.
Specifications
Melting Point | 67°C to 71°C |
Boiling Point | 165°C (10 mmHg) |
Quantity | 25 g |
Beilstein | 607989 |
Sensitivity | Air Sensitive |
Solubility Information | Insoluble in water. Soluble in chlorofrom. |
Formula Weight | 166.18 |
Percent Purity | 98% |
Chemical Name or Material | 2,4-Dimethoxybenzaldehyde |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'