missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethoxy-beta-nitrostyrene, 99%, Thermo Scientific™

Catalog Number 11448947
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1891-10-7
C10H11NO4
209.201
MFCD00015922
NUMXHEUHHRTBQT-AATRIKPKSA-N
2,4-dimethoxy-1-2-nitrovinyl benzene, 2,4-dimethoxy-1-e-2-nitroethenyl benzene, benzene,2,4-dimethoxy-1-2-nitroethenyl, 1-1e-2-nitrovinyl-2,4-dimethoxybenzene, 2,4-dimethoxy-omega-nitrostyrene, benzene, 2,4-dimethoxy-1-2-nitroethenyl, 2,4-dimethoxy-w-nitrostyrene, 2,4-dimethoxy-beta-nitrostyrene, beta-nitro-2,4-dimethoxystyrene, 2-2,4-dimethoxyphenyl nitroethene
688224
2,4-dimethoxy-1-[(E)-2-nitroethenyl]benzene
COC1=CC(=C(C=C1)C=C[N+](=O)[O-])OC
This item is not returnable. View return policy
1891-10-7
C10H11NO4
209.201
MFCD00015922
NUMXHEUHHRTBQT-AATRIKPKSA-N
2,4-dimethoxy-1-2-nitrovinyl benzene, 2,4-dimethoxy-1-e-2-nitroethenyl benzene, benzene,2,4-dimethoxy-1-2-nitroethenyl, 1-1e-2-nitrovinyl-2,4-dimethoxybenzene, 2,4-dimethoxy-omega-nitrostyrene, benzene, 2,4-dimethoxy-1-2-nitroethenyl, 2,4-dimethoxy-w-nitrostyrene, 2,4-dimethoxy-beta-nitrostyrene, beta-nitro-2,4-dimethoxystyrene, 2-2,4-dimethoxyphenyl nitroethene
688224
2,4-dimethoxy-1-[(E)-2-nitroethenyl]benzene
COC1=CC(=C(C=C1)C=C[N+](=O)[O-])OC

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 99°C to 101°C
Quantity 1 g
Formula Weight 209.19
Percent Purity 99%
Chemical Name or Material 2,4-Dimethoxy-beta-nitrostyrene
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'