Learn More
2',4'-Dihydroxypropiophenone, 99%
Product Code. 11451027
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11451027 100 g 100g
11441027 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11451027 Supplier Thermo Scientific Alfa Aesar Supplier No. B23414.22
 Restricted Item
Forventet forsendelse: 02-12-2026
Add to basket
CAS: 5792-36-9 | C9H10O3 | 166.176 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 5792-36-9
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00002280
InChI Key LLBBBYLDTDJMNU-UHFFFAOYSA-N
Synonym 2',4'-dihydroxypropiophenone, 1-2,4-dihydroxyphenyl propan-1-one, 2,4-dihydroxypropiophenone, 4-propionylresorcinol, 1-propanone, 1-2,4-dihydroxyphenyl, respropiophenone, resorcinol fragment, 9, acmc-209m1c, 2',4'-dihydroxy-propiophenone, 2/',4/'-dihydroxypropiophenone
PubChem CID 79856
IUPAC Name 1-(2,4-dihydroxyphenyl)propan-1-one
SMILES CCC(=O)C1=C(C=C(C=C1)O)O
Specifications
Melting Point 98°C to 101°C
Quantity 100 g
Beilstein 1238635
Formula Weight 166.18
Percent Purity 99%
Chemical Name or Material 2',4'-Dihydroxypropiophenone