Få mere at vide
2,4-Dihydroxybenzophenone-d5, TRC
Artikelnummer. 30370275
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30370275 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30370275 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4517831MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2,4-Dihydroxybenzophenone-d5
CAS 91586-06-0
Renhedsgrad noter HPLC
Molekylær formel C13 D5 H5 O3
Mængde 1 mg
Synonym 2,4-Dihydroxybenzophenone D5 (Phenyl D5)
InChI formel InChI=1S/C13H10O3/c14-10-6-7-11(12(15)8-10)13(16)9-4-2-1-3-5-9/h1-8,14-15H/i1D,2D,3D,4D,5D
SMIL [2H]c1c([2H])c([2H])c(C(=O)c2ccc(O)cc2O)c([2H])c1[2H]
IUPAC navn (2,4-dihydroxyphenyl)-(2,3,4,5,6-pentadeuteriophenyl)methanone
Molekylvægt (g/mol) 219.248
Formel vægt 219.094
Procent renhed >95