missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluorophenylacetic acid, 99%, Thermo Scientific Chemicals

Catalog Number p-7025099
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
81228-09-3
C8H5F2O2
171.12
MFCD00009999
QPKZIGHNRLZBCL-UHFFFAOYSA-M
2,4-difluorophenylacetic acid, 2-2,4-difluorophenyl acetic acid, benzeneacetic acid, 2,4-difluoro, 2,4-difluorophenyl acetic acid, 2,4-ddifluorophenylacetic acid, rarechem al bo 0247, 2,4-difluoro-phenyl-acetic acid, 2,4-difluorobenzeneacetic acid, 2,4-difluorophenylaceticacid
123581
2-(2,4-difluorophenyl)acetic acid
[O-]C(=O)CC1=CC=C(F)C=C1F
This item is not returnable. View return policy
81228-09-3
C8H5F2O2
171.12
MFCD00009999
QPKZIGHNRLZBCL-UHFFFAOYSA-M
2,4-difluorophenylacetic acid, 2-2,4-difluorophenyl acetic acid, benzeneacetic acid, 2,4-difluoro, 2,4-difluorophenyl acetic acid, 2,4-ddifluorophenylacetic acid, rarechem al bo 0247, 2,4-difluoro-phenyl-acetic acid, 2,4-difluorobenzeneacetic acid, 2,4-difluorophenylaceticacid
123581
2-(2,4-difluorophenyl)acetic acid
[O-]C(=O)CC1=CC=C(F)C=C1F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 117°C to 119°C
Quantity 5 g
Beilstein 3649727
Formula Weight 172.13
Percent Purity 99%
Chemical Name or Material 2,4-Difluorophenylacetic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'