missing translation for 'onlineSavingsMsg'
Få mere at vide

2,4-Difluorobenzonitrile, TRC

Artikelnummer. 30371623
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30371623 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30371623 Leverandør Toronto Research Chemicals Leverandørnr. TRCD44551525G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale 2,4-Difluorobenzonitrile
CAS 3939-09-1
Molekylær formel C7H3F2N
Mængde 25 g
InChI formel InChI=1S/C7H3F2N/c8-6-2-1-5(4-10)7(9)3-6/h1-3H
SMIL Fc1ccc(C#N)c(F)c1
IUPAC navn 2,4-difluorobenzonitrile
Molekylvægt (g/mol) 139.1
Formel vægt 139.0234
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.