missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluorobenzamide, 97%, Thermo Scientific™

Catalog Number p-7080714
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
85118-02-1
C7H5F2NO
157.12
MFCD00015547
KTXFXDMDYZIXSJ-UHFFFAOYSA-N
benzamide, 2,4-difluoro, difluoro benzamide, pubchem3458, acmc-209q4l, 2,4-difluorobenzamide, 2,4-bis fluoranyl benzamide
123588
2,4-difluorobenzamide
NC(=O)C1=CC=C(F)C=C1F
This item is not returnable. View return policy
85118-02-1
C7H5F2NO
157.12
MFCD00015547
KTXFXDMDYZIXSJ-UHFFFAOYSA-N
benzamide, 2,4-difluoro, difluoro benzamide, pubchem3458, acmc-209q4l, 2,4-difluorobenzamide, 2,4-bis fluoranyl benzamide
123588
2,4-difluorobenzamide
NC(=O)C1=CC=C(F)C=C1F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 160°C to 162°C
Quantity 1 g
Formula Weight 157.12
Percent Purity 97%
Chemical Name or Material 2,4-Difluorobenzamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'