Learn More
2,4-Difluoro-3-methylbenzeneboronic acid pinacol ester, 96%, Thermo Scientific™
Catalog Number 15497956
Click to view available options
Quantity:
250 mg
1 g
1351985-05-1
C13H17BF2O2
254.084
HDAKNAMXPKYUCU-UHFFFAOYSA-N
2,4-difluoro-3-methylphenylboronic acid pinacol ester, 2,4-difluoro-3-methylbenzene boronic acid pinacol ester, 2-2,4-difluoro-3-methylphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
100913465
2-(2,4-difluoro-3-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C2=C(C(=C(C=C2)F)C)F
1351985-05-1
C13H17BF2O2
254.084
HDAKNAMXPKYUCU-UHFFFAOYSA-N
2,4-difluoro-3-methylphenylboronic acid pinacol ester, 2,4-difluoro-3-methylbenzene boronic acid pinacol ester, 2-2,4-difluoro-3-methylphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
100913465
2-(2,4-difluoro-3-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C2=C(C(=C(C=C2)F)C)F
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 254.08
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 2,4-Difluoro-3-methylbenzeneboronic acid pinacol ester