Få mere at vide
2,4-Dicyano-3-(2-methylpropyl)-pentanediamide (Mixture of Diastereomers), TRC
Artikelnummer. 30371114
Skift visning
Mængde:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30371114 1 g
30369433 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30371114 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4377801G
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 185815-56-9
Molekylær formel C11H16N4O2
Molekylvægt (g/mol) 236.27
Synonym Pregabalin Impurity 9
SMIL NC(C(C#N)C(CC(C)C)C(C(N)=O)C#N)=O
Tekniske data
Kemisk navn eller materiale 2,4-Dicyano-3-(2-methylpropyl)-pentanediamide (Mixture of Diastereomers)
Mængde 1 g
InChI formel InChI=1S/C11H16N4O2/c1-6(2)3-7(8(4-12)10(14)16)9(5-13)11(15)17/h6-9H,3H2,1-2H3,(H2,14,16)(H2,15,17)
Formel vægt 236.1273