Få mere at vide
2,4-Dichlorothiazole-5-carboxaldehyde, 96%, Thermo Scientific™
Artikelnummer. 11355499
Skift visning
Mængde:
250 mg
1 g
Pakningsstørrelse:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
11355499 250 mg 250mg
11365499 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 11355499 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. H50456.MD
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 92972-48-0
Molekylær formel C4HCl2NOS
Molekylvægt (g/mol) 182.018
MDL nummer MFCD00793013
InChI nøgle CFEKBKCGPASOFI-UHFFFAOYSA-N
Synonym 2,4-dichloro-5-thiazolecarboxaldehyde, 2,4-dichlorothiazole-5-carbaldehyde, 2,4-dichloro-5-formylthiazole, 2,4-dichlorothiazole-5-carboxaldehyde, 5-thiazolecarboxaldehyde, 2,4-dichloro, 2,4-dichloro-5-formyl-1,3-thiazole, 2,4-dichloro-5-thiazole-carboxaldehyde, 2,4-dichlorothiazol-5-carboxaldehyde, 2,4-dichloro-thiazole-5-carbaldehyde, pubchem18196
PubChem CID 1488672
IUPAC navn 2,4-dichloro-1,3-thiazole-5-carbaldehyde
SMIL C(=O)C1=C(N=C(S1)Cl)Cl
Tekniske data
Kemisk navn eller materiale 2,4-Dichlorothiazole-5-carboxaldehyde
Smeltepunkt 48°C to 49°C
Mængde 250 mg
Formel vægt 182.03
Procent renhed 96%