Få mere at vide
(2,4-Dichlorophenoxy-d3)acetic-d2 Acid, CDN
Artikelnummer. 30318873
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30318873 0.005 g
30328093 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30318873 Leverandør CDN Leverandørnr. CDND51210.005G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2,4-D D5
CAS 352438-69-8
Molekylær formel C8 D5 H Cl2 O3
Mængde 0.005 g
Synonym Acetic-2,2-d2 acid, 2-(4,6-dichlorophenoxy-2,3,5-d3)- (ACI), 2-(4,6-Dichlorophenoxy-2,3,5-d3)acetic-2,2-d2 acid (ACI), Acetic-d2 acid, (4,6-dichlorophenoxy-2,3,5-d3)- (9 CI), 2,4-D D5, 2,4-D-d5, (2,4-Dichloro-3,5,6-trideuteriophenoxy)dideuterioacetic Acid, (2,4-Dichlorophenoxy-d3)acetic-d2 Acid
InChI formel InChI=1 S/C8H6Cl2O3/c9-5-1-2-7(6(10)3-5)13-4-8(11)12/h1-3 H,4H2,(H,11,12)/i1D,2 D,3 D,4D2
SMIL [2 H]c1c([2 H])c(OC([2 H])([2 H])C(=O)O)c(Cl)c([2 H])c1Cl
IUPAC navn 2,2-dideuterio-2-(2,4-dichloro-3,5,6-trideuteriophenoxy)acetic acid
Molekylvægt (g/mol) 226.0683
Formel vægt 225.0008 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity