Få mere at vide
2,4-Dichlorophenol-13C6, TRC
Artikelnummer. 30135602
Skift visning
Mængde:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30135602 25 mg
30137833 10 mg
30005153 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30135602 Leverandør Toronto Research Chemicals Leverandørnr. TRCD43568625MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1202864-83-2
Molekylær formel 13C6 H4 Cl2 O
Molekylvægt (g/mol) 168.96
Synonym 2,4-Dichlorophenol 13C6, 2,4-Dichlorophenol-1,2,3,4,5,6-13C6
IUPAC navn 2,4-dichloro(1,2,3,4,5,6-^{13}C_{6})cyclohexa-1,3,5-trien-1-ol
SMIL O[13c]1[13cH][13cH][13c](Cl)[13cH][13c]1Cl
Tekniske data
Kemisk navn eller materiale 2,4-Dichlorophenol13C6
Mængde 25 mg
Renhedsgrad noter GC
InChI formel InChI=1S/C6H4Cl2O/c7-4-1-2-6(9)5(8)3-4/h1-3,9H/i1+1,2+1,3+1,4+1,5+1,6+1
Formel vægt 167.984
Procent renhed >95%