Få mere at vide
2,4'-Dichlorodiphenyltrichloroethane-d8, TRC
Artikelnummer. 30240683
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30240683 10 mg
30240671 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30240683 Leverandør Toronto Research Chemicals Leverandørnr. TRCD43420710MG
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 221899-88-3
Molekylær formel C14 2H8 H Cl5
Molekylvægt (g/mol) 362.54
Synonym 2,4'-DDT D8 (1,1,1-Trichloro-2-(p-chlorophenyl)-2-(o-chlorophenyl)ethane D8 (ring D8)), Benzene-1,2,3,4-d4, 5-chloro-6-[2,2,2-trichloro-1-(4-chlorophenyl-2,3,5,6-d4)ethyl]- (9CI)
IUPAC navn 1-chloro-2,3,4,5-tetradeuterio-6-[2,2,2-trichloro-1-(4-chloro-2,3,5,6-tetradeuteriophenyl)ethyl]benzene
SMIL [2H]c1c([2H])c([2H])c(C(c2c([2H])c([2H])c(Cl)c([2H])c2[2H])C(Cl)(Cl)Cl)c(Cl)c1[2H]
Tekniske data
Kemisk navn eller materiale 2,4'-DDT D8
Mængde 10 mg
InChI formel InChI=1S/C14H9Cl5/c15-10-7-5-9(6-8-10)13(14(17,18)19)11-3-1-2-4-12(11)16/h1-8,13H/i1D,2D,3D,4D,5D,6D,7D,8D
Formel vægt 359.9649