Få mere at vide
2,4-Dichloro-6-nitroaniline, TRC
Artikelnummer. 30383594
Skift visning
Mængde:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30383594 500 mg
30378061 250 mg
30379143 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30383594 Leverandør Toronto Research Chemicals Leverandørnr. TRCD435165500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2683-43-4
Molekylær formel C6H4Cl2N2O2
Molekylvægt (g/mol) 207.01
IUPAC navn 2,4-dichloro-6-nitroaniline
SMIL Nc1c(Cl)cc(Cl)cc1[N+](=O)[O-]
Tekniske data
Kemisk navn eller materiale 2,4-Dichloro-6-nitroaniline
Mængde 500 mg
InChI formel InChI=1S/C6H4Cl2N2O2/c7-3-1-4(8)6(9)5(2-3)10(11)12/h1-2H,9H2
Formel vægt 205.965