missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-6-methylpyridine, 97%, Thermo Scientific™

Catalog Number p-7077434
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
42779-56-6
C6H5Cl2N
162.02
WUGTXQVNSRFDNV-UHFFFAOYSA-N
2,4-dichloro-6-picoline, 2,4-dichloro-6-methyl-pyridine, pubchem5465, d0ri6l, 2-methyl-4,6-dichloropyridine, pyridine,2,4-dichloro-6-methyl
11959083
2,4-dichloro-6-methylpyridine
CC1=NC(=CC(=C1)Cl)Cl
This item is not returnable. View return policy
42779-56-6
C6H5Cl2N
162.02
WUGTXQVNSRFDNV-UHFFFAOYSA-N
2,4-dichloro-6-picoline, 2,4-dichloro-6-methyl-pyridine, pubchem5465, d0ri6l, 2-methyl-4,6-dichloropyridine, pyridine,2,4-dichloro-6-methyl
11959083
2,4-dichloro-6-methylpyridine
CC1=NC(=CC(=C1)Cl)Cl
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 162.02
Percent Purity 97%
Chemical Name or Material 2,4-Dichloro-6-methylpyridine
compliance-icons
Product Identifier
  • 2,4-Dichloro-6-methylpyridine, 97%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'