missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-6-methylphenol, 98%, Thermo Scientific Chemicals

Catalog Number 15481459
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
1570-65-6
C7H6Cl2O
177.024
MFCD00019988
WJQZZLQMLJPKQH-UHFFFAOYSA-N
15292
2,4-dichloro-6-methylphenol
CC1=CC(=CC(=C1O)Cl)Cl
This item is not returnable. View return policy
1570-65-6
C7H6Cl2O
177.024
MFCD00019988
WJQZZLQMLJPKQH-UHFFFAOYSA-N
15292
2,4-dichloro-6-methylphenol
CC1=CC(=CC(=C1O)Cl)Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 52°C to 56°C
Color Brown
Boiling Point 225°C (literature)
Quantity 250 mg
UN Number UN3077
Beilstein 1864865
Synonym 4,6-dichloro-o-cresol, 4,6-dichloro-2-methylphenol, phenol, 2,4-dichloro-6-methyl, o-cresol, 4,6-dichloro, 4,6-dichlorocresol, phenol,2,4-dichloro-6-methyl, dichlorocresol, o-cresol,6-dichloro, 4,6-dichlor-o-kresol, acmc-1c0mk
Formula Weight 177.02
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2,4-Dichloro-6-methylphenol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'