Få mere at vide
2,4-Dichloro-5-nitrophenyl Isopropyl Ether, TRC
Artikelnummer. 30372185
Skift visning
Mængde:
500 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30372185 500 mg
30370970 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30372185 Leverandør Toronto Research Chemicals Leverandørnr. TRCD435550500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 41200-97-9
Molekylær formel C9H9Cl2NO3
Molekylvægt (g/mol) 250.08
Synonym 1,5-Dichloro-2-(1-methylethoxy)-4-nitrobenzene
SMIL CC(C)Oc1cc(c(Cl)cc1Cl)[N+](=O)[O-]
Tekniske data
Kemisk navn eller materiale 2,4-Dichloro-5-nitrophenyl Isopropyl Ether
Mængde 500 mg
InChI formel InChI=1S/C9H9Cl2NO3/c1-5(2)15-9-4-8(12(13)14)6(10)3-7(9)11/h3-5H,1-2H3
Formel vægt 248.9959