missing translation for 'onlineSavingsMsg'
Få mere at vide

2,4-Dibromobutyric Acid, TRC

Artikelnummer. 30377296
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30377296 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30377296 Leverandør Toronto Research Chemicals Leverandørnr. TRCD42518850MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale 2,4-Dibromobutyric Acid
CAS 63164-16-9
Molekylær formel C4 H6 Br2 O2
Mængde 50 mg
InChI formel InChI=1S/C4H6Br2O2/c5-2-1-3(6)4(7)8/h3H,1-2H2,(H,7,8)
SMIL OC(=O)C(Br)CCBr
IUPAC navn 2,4-dibromobutanoic acid
Molekylvægt (g/mol) 245.897
Formel vægt 243.8735
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.