Få mere at vide
2,4-Dibromo-6-Fluoro-Benzonitrile, TRC
Artikelnummer. 30387856
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30387856 100 mg
30377252 10 mg
30385856 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30387856 Leverandør Toronto Research Chemicals Leverandørnr. TRCD472390100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1393585-66-4
Molekylær formel C7H2NFBr2
Molekylvægt (g/mol) 278.9
IUPAC navn 2,4-dibromo-6-fluorobenzonitrile
SMIL Fc1cc(Br)cc(Br)c1C#N
Tekniske data
Kemisk navn eller materiale 2,4-Dibromo-6-Fluoro-Benzonitrile
Mængde 100 mg
InChI formel InChI=1S/C7H2Br2FN/c8-4-1-6(9)5(3-11)7(10)2-4/h1-2H
Formel vægt 276.8538