missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Dibromo-3-pentanone, mixture of stereoisomers, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
2,4-Dibromo-2,4-dimethyl-3-pentanone may be used in the synthesis of 2-dimethylamino-4-methylene-1,3-dioxolanes, via debromination using zinc-copper couple in dimethylformamide and dimethylacetamide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,4-Dibromo-2,4-dimethyl-3-pentanone may be used in the synthesis of 2-dimethylamino-4-methylene-1,3-dioxolanes, via debromination using zinc-copper couple in dimethylformamide and dimethylacetamide.
Solubility
Slightly soluble in water(50 g/L (20°C).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, reducing agents, bases.
2,4-Dibromo-2,4-dimethyl-3-pentanone may be used in the synthesis of 2-dimethylamino-4-methylene-1,3-dioxolanes, via debromination using zinc-copper couple in dimethylformamide and dimethylacetamide.
Solubility
Slightly soluble in water(50 g/L (20°C).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, reducing agents, bases.
Specifications
Boiling Point | 67°C to 82°C (10 mmHg) |
Odor | Pungent |
Quantity | 10 g |
UN Number | UN3265 |
Solubility Information | Slightly soluble in water(50g/L (20°C). |
Formula Weight | 243.91 |
Percent Purity | 97% |
Chemical Name or Material | 2,4-Dibromo-3-pentanone, mixture of stereoisomers |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'