Få mere at vide
2,4-Diaminotoluene-d3, TRC
Artikelnummer. 30358985
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30358985 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30358985 Leverandør Toronto Research Chemicals Leverandørnr. TRCD41678250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2,4-Diaminotoluene-d3
CAS 71111-08-5
Molekylær formel C7 2H3 H7 N2
Mængde 50 mg
Synonym 1,3-Benzenediamine, 4-(methyl-d3)- (9CI), 4-(Methyl-d3)-1,3-benzenediamine, 2,4-Diaminotoluene-d3, 2,4-Tolylenediamine-d3, 3-Amino-4-(methyl-d3)aniline, 4-(Methyl-d3)-1,3-diaminobenzene, 4-(Methyl-d3)-m-phenylenediamine, m-Toluenediamine-d3, 2,4-Diaminotoluene-D3 (methyl-D3)
InChI formel InChI=1S/C7H10N2/c1-5-2-3-6(8)4-7(5)9/h2-4H,8-9H2,1H3/i1D3
SMIL [2H]C([2H])([2H])c1ccc(N)cc1N
IUPAC navn 4-(trideuteriomethyl)benzene-1,3-diamine
Molekylvægt (g/mol) 125.19
Formel vægt 125.1032