Få mere at vide
2,4-Diaminotoluene-alpha,alpha,alpha-d3, CDN
Artikelnummer. 30317985
Skift visning
Mængde:
0.05 g
0.1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30317985 0.05 g
30325642 0.1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30317985 Leverandør CDN Leverandørnr. CDND54720.05G
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2,4-Diaminotoluene-d3
CAS 71111-08-5
Molekylær formel C7 2H3 H7 N2
Mængde 0.05 g
InChI formel InChI=1 S/C7H10N2/c1-5-2-3-6(8)4-7(5)9/h2-4 H,8-9H2,1H3/i1D3
SMIL [2 H]C([2 H])([2 H])c1ccc(N)cc1N
IUPAC navn 4-(trideuteriomethyl)benzene-1,3-diamine
Molekylvægt (g/mol) 125.19
Formel vægt 125.1032 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity