Få mere at vide
(2,4-Diaminopteridin-6-yl)methanol Hydrochloride, Mikromol™
Artikelnummer. 17904106
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17904106 25 mg
17951176 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17904106 Leverandør Mikromol Leverandørnr. MM0040.080025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(2,4-Diaminopteridin-6-yl)methanol Hydrochloride
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (2,4-Diaminopteridin-6-yl)methanol Hydrochloride
CAS 73978-41-3
Analyt- eller komponentnavne (2,4-Diaminopteridin-6-yl)methanol Hydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C7 H8 N6 O . Cl H
Certificeringer/compliance ISO 9001
Synonym (2,4-Diaminopteridin-6-yl)methanol Hydrochloride, Methotrexate Imp. A (EP) as Hydrochloride, Methotrexate Impurity A as Hydrochloride
InChI formel InChI=1S/C7H8N6O.ClH/c8-5-4-6(13-7(9)12-5)10-1-3(2-14)11-4;/h1,14H,2H2,(H4,8,9,10,12,13);1H
SMIL Cl.Nc1nc(N)c2nc(CO)cnc2n1
IUPAC navn (2,4-diaminopteridin-6-yl)methanol;hydrochloride
Molekylvægt (g/mol) 228.64
Mængde 25 mg
Formel vægt 228.0526 g/mol