Få mere at vide
2,4-Diamino-7-Pteridinemethanol, TRC
Artikelnummer. 30135520
Skift visning
Mængde:
25 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30135520 250 mg
30003511 25 mg
30008740 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30135520 Leverandør Toronto Research Chemicals Leverandørnr. TRCP881330250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 881-33-4
Molekylær formel C7H8N6O
Molekylvægt (g/mol) 192.18
Synonym (2,4-Diaminopteridin-7-yl)methanol
IUPAC navn (2,4-diaminopteridin-7-yl)methanol
SMIL Nc1nc(N)c2ncc(CO)nc2n1
Tekniske data
Kemisk navn eller materiale 2,4-Diamino-7-Pteridinemethanol
Mængde 250 mg
InChI formel InChI=1S/C7H8N6O/c8-5-4-6(13-7(9)12-5)11-3(2-14)1-10-4/h1,14H,2H2,(H4,8,9,11,12,13)
Formel vægt 192.076