Få mere at vide
2,4-Diamino-6-methyl-1,3,5-triazine, 96%, Thermo Scientific™
Artikelnummer. 11450218
Skift visning
Mængde:
100 g
500 g
Pakningsstørrelse:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
11450218 500 g 500g
11440218 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 11450218 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. B23768.36
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 542-02-9
Molekylær formel C4H7N5
Molekylvægt (g/mol) 125.135
MDL nummer MFCD00023192
InChI nøgle NJYZCEFQAIUHSD-UHFFFAOYSA-N
Synonym acetoguanamine, 2,4-diamino-6-methyl-1,3,5-triazine, 1,3,5-triazine-2,4-diamine, 6-methyl, 2-methyl-4,6-diamino-1,3,5-triazine, 2,4-diamino-6-methyl-s-triazine, 6-methyl-1,3,5-triazine-2,4-diyldiamine, s-triazine, 2,4-diamino-6-methyl, acetoguanamin, ccris 8909, dsstox_cid_9186
PubChem CID 10949
ChEBI CHEBI:72475
IUPAC navn 6-methyl-1,3,5-triazine-2,4-diamine
SMIL CC1=NC(=NC(=N1)N)N
Tekniske data
Kemisk navn eller materiale 2,4-Diamino-6-methyl-1,3,5-triazine
Smeltepunkt ∼280°C (decomposition)
Mængde 500 g
Beilstein 118348
Formel vægt 125.13
Procent renhed 96%