Få mere at vide
2,4-Diamino-6-(hydroxymethyl)pteridine Hydrochloride, TRC
Artikelnummer. 30367550
Skift visning
Mængde:
1 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30367550 1 g
30350042 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30367550 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4163951G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 73978-41-3
Molekylær formel C7 H8 N6 O . Cl H
Molekylvægt (g/mol) 228.64
Synonym (2,4-Diaminopteridin-6-yl)methanol Hydrochloride, Methotrexate Imp. A (EP) as Hydrochloride
IUPAC navn (2,4-diaminopteridin-6-yl)methanol;hydrochloride
SMIL Cl.Nc1nc(N)c2nc(CO)cnc2n1
Tekniske data
Kemisk navn eller materiale (2,4-Diaminopteridin-6-yl)methanol Hydrochloride
Mængde 1 g
InChI formel InChI=1S/C7H8N6O.ClH/c8-5-4-6(13-7(9)12-5)10-1-3(2-14)11-4;/h1,14H,2H2,(H4,8,9,10,12,13);1H
Formel vægt 228.0526