missing translation for 'onlineSavingsMsg'
Få mere at vide

2,4-Di-tert-butylphenol-d19 (Major), TRC

Product Code. 30369258
Change view
Click to view available options
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30369258 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30369258 Supplier Toronto Research Chemicals Supplier No. TRCD42947210MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 2,4-Di-tert-butylphenol-d19
CAS 1577233-55-6
Molekylær formel C14H3D19O
Mængde 10 mg
Synonym 4,6-Bis[1,1-di(methyl-d3)ethyl-2,2,2-d3]phen-2-d-ol
InChI formel InChI=1S/C14H22O/c1-13(2,3)10-7-8-12(15)11(9-10)14(4,5)6/h7-9,15H,1-6H3/i1D3,2D3,3D3,4D3,5D3,6D3,8D
SMIL OC1=C(C(C([2H])([2H])[2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])C=C(C(C([2H])([2H])[2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])C=C1[2H]
Molekylvægt (g/mol) 225.44
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.