Få mere at vide
2-[[(4-Cyanophenyl)amino]methyl]-1-methyl-1H-benzimidazole-5-carboxylic-d3 Acid Ethyl Ester, TRC
Artikelnummer. 30367488
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30367488 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30367488 Leverandør Toronto Research Chemicals Leverandørnr. TRCC9821325MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-(((4-Cyanophenyl)amino)methyl)-1-methyl-1H-benzimidazole-5-carboxylic-d3 Acid Ethyl Ester
CAS 1346601-47-5
Molekylær formel C19H15D3N4O2
Mængde 5 mg
InChI formel InChI=1S/C19H18N4O2/c1-3-25-19(24)14-6-9-17-16(10-14)22-18(23(17)2)12-21-15-7-4-13(11-20)5-8-15/h4-10,21H,3,12H2,1-2H3/i2D3
SMIL [2H]C([2H])([2H])n1c(CNc2ccc(cc2)C#N)nc3cc(ccc13)C(=O)OCC
IUPAC navn ethyl 2-[(4-cyanoanilino)methyl]-1-(trideuteriomethyl)benzimidazole-5-carboxylate
Molekylvægt (g/mol) 337.39
Formel vægt 337.1618