missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Chlorophenyl)-1-phenylethanone, TRC

Product Code. 30362259
Change view
Click to view available options
Mængde:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30362259 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30362259 Supplier Toronto Research Chemicals Supplier No. TRCC3778651G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 2-(4-Chlorophenyl)-1-phenylethanone
CAS 6332-83-8
Molekylær formel C14H11ClO
Mængde 1 g
Synonym -(4-Chlorophenyl)-1-phenyl-1-ethanone, 2-(p-Chlorophenyl)acetophenone, 4-Chlorobenzyl Phenyl Ketone, NSC 38739, p-Chlorobenzyl Phenyl Ketone
InChI formel InChI=1S/C14H11ClO/c15-13-8-6-11(7-9-13)10-14(16)12-4-2-1-3-5-12/h1-9H,10H2
SMIL O=C(CC1=CC=C(C=C1)Cl)C2=CC=CC=C2
IUPAC navn 2-(4-chlorophenyl)-1-phenylethan-1-one
Molekylvægt (g/mol) 230.69
Formel vægt 230.05
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.