missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Chlorophenoxy)nicotinic acid, 98%, Thermo Scientific™

Catalog Number 11496927
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
51362-37-9
C12H8ClNO3
249.65
MFCD00052116
FVAZJGJEENAKRJ-UHFFFAOYSA-N
2-4-chlorophenoxy nicotinic acid, 2-4-chlorophenoxy pyridine-3-carboxylic acid, 2-4-chloro-phenoxy-nicotinic acid, 3-pyridinecarboxylic acid, 2-4-chlorophenoxy, acmc-1avkg, maybridge1_001700, 3-pyridinecarboxylicacid, 2-4-chlorophenoxy
609454
2-(4-chlorophenoxy)pyridine-3-carboxylic acid
C1=CC(=C(N=C1)OC2=CC=C(C=C2)Cl)C(=O)O
This item is not returnable. View return policy
51362-37-9
C12H8ClNO3
249.65
MFCD00052116
FVAZJGJEENAKRJ-UHFFFAOYSA-N
2-4-chlorophenoxy nicotinic acid, 2-4-chlorophenoxy pyridine-3-carboxylic acid, 2-4-chloro-phenoxy-nicotinic acid, 3-pyridinecarboxylic acid, 2-4-chlorophenoxy, acmc-1avkg, maybridge1_001700, 3-pyridinecarboxylicacid, 2-4-chlorophenoxy
609454
2-(4-chlorophenoxy)pyridine-3-carboxylic acid
C1=CC(=C(N=C1)OC2=CC=C(C=C2)Cl)C(=O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 157°C to 159°C
Quantity 1 g
Formula Weight 249.64
Percent Purity 98%
Chemical Name or Material 2-(4-Chlorophenoxy)nicotinic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'