Få mere at vide
2-(4-Bromophenoxy)-N,N-dimethylethylamine-d6, TRC
Artikelnummer. 30348433
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30348433 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30348433 Leverandør Toronto Research Chemicals Leverandørnr. TRCB68638250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-(4-Bromophenoxy)-N,N-dimethylethylamine-d6
CAS 1794787-00-0
Molekylær formel C10H8D6BrNO
Mængde 50 mg
Synonym 1-Bromo-4-[2-(dimethylamino)ethoxy]benzene-d6, 4-Bromophenyl 2-(Dimethylamino)ethyl Ether-d6, N,N-Dimethyl-2-(p-bromophenoxy)ethylamine-d6
InChI formel InChI=1S/C10H14BrNO/c1-12(2)7-8-13-10-5-3-9(11)4-6-10/h3-6H,7-8H2,1-2H3/i1D3,2D3
SMIL [2H]C([2H])([2H])N(CCOc1ccc(Br)cc1)C([2H])([2H])[2H]
IUPAC navn 2-(4-bromophenoxy)-N,N-bis(trideuteriomethyl)ethanamine
Molekylvægt (g/mol) 250.17
Formel vægt 249.0635