Learn More
2-(4-Bromobutyl)isoindoline-1,3-dione, 97%, BLD Pharm™
Product Code. 31007128
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
100 g
500 g
1 kg
Unit Size:
100g
10g
1kg
25g
500g
5g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31007128 5 g 5g
31002449 500 g 500g
30990112 25 g 25g
31004463 100 g 100g
31005948 10 g 10g
30990347 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 31007128 Supplier BLD Pharm Supplier No. BD138505g
 Restricted Item
Forventet forsendelse: 27-11-2026
Specifications
CAS 5394-18-3
Molecular Formula C12H12BrNO2
MDL Number MFCD00005905
Quantity 5 g
SMILES C1=CC=CC2=C1C(N(CCCCBr)C2=O)=O
Molecular Weight (g/mol) 282.13
Percent Purity 97%
Chemical Name or Material 2-(4-Bromobutyl)isoindoline-1,3-dione