Få mere at vide
2-(4-Boc-piperazino)pyrimidine-5-boronic acid pinacol ester, TRC
Artikelnummer. 30346782
Skift visning
Mængde:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30346782 100 mg
30335966 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30346782 Leverandør Toronto Research Chemicals Leverandørnr. TRCB663205100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-(4-Boc-piperazino)pyrimidine-5-boronic acid pinacol ester
CAS 940284-98-0
Molekylær formel C19H31BN4O4
Mængde 100 mg
InChI formel InChI=1S/C19H31BN4O4/c1-17(2,3)26-16(25)24-10-8-23(9-11-24)15-21-12-14(13-22-15)20-27-18(4,5)19(6,7)28-20/h12-13H,8-11H2,1-7H3
SMIL CC(C)(C)OC(=O)N1CCN(CC1)c2ncc(cn2)B3OC(C)(C)C(C)(C)O3
IUPAC navn tert-butyl 4-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-yl]piperazine-1-carboxylate
Molekylvægt (g/mol) 390.29
Formel vægt 390.2438